Order Entry
Switzerland
ContactUsLinkComponent
Anti-Caspase 9 Mouse Monoclonal Antibody [clone: LAP6 96-2-22]
Anti-Caspase 9 Mouse Monoclonal Antibody [clone: LAP6 96-2-22]
Catalog # GNTXGTX30245
Supplier:  Genetex
Anti-Caspase 9 Mouse Monoclonal Antibody [clone: LAP6 96-2-22]
Catalog # GNTXGTX30245
Supplier:  Genetex

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Caspase 9
  • Antigen symbol:
    CASP9
  • Clonality:
    Monoclonal
  • Clone:
    LAP6 96-2-22
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Format:
    Ascites fluid
  • Storage temperature:
    –20 °C
  • Immunogen:
    Recombinant human Caspase 9 pro-domain protein.
  • Purification:
    Ascitic fluid
  • Pk:
    200 µl

Specifications

About this item

Type: Primary
Antigen: Caspase 9
Clonality: Monoclonal
Clone: [LAP6 96-2-22]
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human