Anti-TNF alpha Mouse Monoclonal Antibody [clone: F6C5]
GNTXGTX28349
: Genetex
- Antibody type:Primary
- Antigen symbol:TNF alpha
- Clonality:Monoclonal
- Clone:F6C5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Storage temperature:+4 °C
- Immunogen:Human recombinant tumor necrosis factor of alpha type (TNF alpha)
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: TNF alpha
Clonality: Monoclonal
Clone: [F6C5]
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human