Anti-TNF alpha Mouse Monoclonal Antibody [clone: 2C8]
Catalog # GNTXGTX28348
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen symbol:TNF alpha
- Clonality:Monoclonal
- Clone:2C8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Storage temperature:+4 °C
- Immunogen:Human recombinant tumor necrosis factor of alpha type.
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: TNF alpha
Clonality: Monoclonal
Clone: [2C8]
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human