Auftragserfassung
Switzerland
ContactUsLinkComponent
Anti-MUC1 Mouse Monoclonal Antibody [clone: M2C5]
Anti-MUC1 Mouse Monoclonal Antibody [clone: M2C5]
Catalog # GNTXGTX28323
Supplier:  Genetex
Anti-MUC1 Mouse Monoclonal Antibody [clone: M2C5]
Catalog # GNTXGTX28323
Supplier:  Genetex

Specifications

  • Antikörper-Typ:
    Primär
  • Antigen-Symbol:
    MUC1
  • Klonalität:
    Monoklonal
  • Klon:
    M2C5
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Wirt:
    Mouse
  • ImmunoChemie:
    Yes
  • Isotyp:
    IgG
  • Reaktivität:
    Human
  • Molecular weight:
    300 kDa
  • Lagertemperatur:
    +4 °C
  • Immunogen:
    High molecular weight (more than 300 kDa) glycoprotein from human milk-fat globule molecule
  • Reinigung:
    purified by Protein A chromatography
  • Größe:
    100 µg
  • VE:
    100 µG

Specifications

About this item

Type: Primary
Antigen: MUC1
Clonality: Monoclonal
Clone: [M2C5]
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human