Anti-Cardiac Troponin T Mouse Monoclonal Antibody [clone: 1C11]
Catalog # GNTXGTX28295
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Cardiac Troponin T
- Antigen symbol:cTnT
- Clonality:Monoclonal
- Clone:1C11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Dog
- Western blot:Yes
- Storage temperature:+4 °C
- Immunogen:Full length native protein (purified) (Human).
- Purification:purified by Protein A chromatography
- Size:200 µg
- Pk:200 µG
Specifications
About this item
Type: Primary
Antigen: Cardiac Troponin T
Clonality: Monoclonal
Clone: [1C11]
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog