Anti-alpha 1 FETO Receptor Mouse Monoclonal Antibody [clone: 50]
Catalog # GNTXGTX28276
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:alpha-1-Fetoprotein Receptor
- Clonality:Monoclonal
- Clone:5E1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Human
- Antigen synonyms:AFP Receptor
- Storage temperature:+4 °C
- Immunogen:Full length native protein (purified) (Human).
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: alpha 1 FETO Receptor
Clonality: Monoclonal
Clone: [5E1]
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human