Anti-Fibrinogen Mouse Monoclonal Antibody [clone: 49D2]
Catalog # GNTXGTX14801
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Fibrinogen
- Clonality:Monoclonal
- Clone:49D2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Storage temperature:+4 °C
- Immunogen:fibrinopeptide A conjugated with a carrier protein.
- Purification:purified by Protein A chromatography
- Size:200 µg
- Pk:200 µG
Specifications
About this item
Type: Primary
Antigen: Fibrinogen
Clonality: Monoclonal
Clone: [49D2]
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human