Anti-LAP2 Mouse Monoclonal Antibody [clone: 6E10]
Catalog # GNTXGTX11823
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen symbol:LAP2
- Clonality:Monoclonal
- Clone:6E10
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Bovine
- Western blot:Yes
- Amino acid number:29 to 50
- Storage temperature:–20 °C
- Immunogen:Synthetic peptide: GEQRKDVYVQLYLQHLTARNRP, corresponding to amino acids 29-50 of Human LAP2, common to all isoforms.
- Purification:purified by Protein A chromatography
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: LAP2
Clonality: Monoclonal
Clone: 6E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Cow