Anti-IL12 Mouse Monoclonal Antibody [clone: 1-1A4]
Catalog # GNTXGTX11446
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen symbol:IL12
- Clonality:Monoclonal
- Clone:1-1A4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Storage temperature:–20 °C
- Immunogen:Recombinant full length protein (Human).
- Purification:purified by Protein G chromatography
- Size:250 µg
- Pk:250 µG
Specifications
About this item
Type: Primary
Antigen: IL12
Clonality: Monoclonal
Clone: [1-1A4]
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human