Anti-ROCK1 Rabbit Polyclonal Antibody
Catalog # GNTXGTX113266
Anbieter: Genetex
Technische Daten
- Antibody type:Primary
- Antigen symbol:ROCK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Amino acid number:corresponding to a region within amino acids 107 and 371
- Molecular weight:158 kDa
- Storage temperature:–20 °C
- Immunogen:Recombinant protein fragment contain a sequence corresponding to a region within amino acids 107 and 371 (Q13464) of ROCK1
- Purification:purified by antigen affinity chromatography
- Pk:100 µl
Technische Daten
Über diesen Artikel
Type: Primary
Antigen: ROCK1
Clonality: Polyclonal
Clone:
Conjugation: No
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human