Anti-Prolactin Mouse Monoclonal Antibody [clone: INN-hPRL-1]
Catalog # GNTXGTX11301
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen symbol:Prolactin
- Clonality:Monoclonal
- Clone:INN-hPRL-1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Storage temperature:–20 °C
- Immunogen:Unfortunately, this information is considered to be commercially sensitive.
- Purification:purified
- Size:250 µg
- Pk:250 µG
Specifications
About this item
Type: Primary
Antigen: Prolactin
Clonality: Monoclonal
Clone: [INN-hPRL-1]
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human