Anti-Cardiac Troponin T Mouse Monoclonal Antibody [clone: 1A11]
Katalognr. GNTXGTX10213
Anbieter: Genetex
Technische Daten
- Antibody type:Primary
- Antigen name:Cardiac Troponin T
- Antigen symbol:cTnT
- Clonality:Monoclonal
- Clone:1A11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Cat
- Western blot:Yes
- Storage temperature:+4 °C
- Immunogen:Full length protein (Human).
- Purification:purified by Protein A chromatography
- Size:200 µg
- Pk:200 µG
Technische Daten
Über diesen Artikel
Type: Primary
Antigen: Cardiac Troponin T
Clonality: Monoclonal
Clone: [1A11]
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Cat