Anti-COI1 Rabbit Polyclonal Antibody
Catalog # AGRIAS122637
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:COI1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- UniProtKB:O04197
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide derived from Arabidopsis thaliana COI1 protein,
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: COI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human