Anti-TIP2 Rabbit Polyclonal Antibody
Catalog # AGRIAS122619
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:TIP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:A. thaliana
- Western blot:Yes
- Cross adsorption:No
- UniProtKB:Q9FGL2
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide conserved in Arabidopsis thaliana TIP2;1 (100 %)
- Size:200 µg
- Pk:200 µG
Specifications
About this item
Type: Primary
Antigen: TIP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana