Anti-FMR Rabbit Polyclonal Antibody
Catalog # AGRIAS122618
Anbieter: Agrisera
Technische Daten
- Antibody type:Primary
- Antigen symbol:FMR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide derived Chlamydomonas reinhardtii FMR protein seqeunce, protein ID 145357.
- Size:200 µg
- Pk:200 µG
Technische Daten
Über diesen Artikel
Type: Primary
Antigen: FMR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: