Anti-AHK2 Rabbit Polyclonal Antibody
Catalog # AGRIAS122113
Anbieter: Agrisera
Technische Daten
- Antibody type:Primary
- Antigen symbol:AHK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Rat
- Western blot:Yes
- Cross adsorption:No
- UniProtKB:Q9C5U2
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide derived from Arabidopsis thaliana AHK2 protein sequence, TAIR: AT5G35750,
- Size:50 µg
- Pk:50 µG
Technische Daten
Über diesen Artikel
Type: Primary
Antigen: AHK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat