Order Entry
Switzerland
ContactUsLinkComponent
Anti-IgG (H+L) Donkey Polyclonal Antibody (DyLight® 594)
Anti-IgG (H+L) Donkey Polyclonal Antibody (DyLight® 594)
Catalog # AGRIAS122090
Supplier:  Agrisera
Anti-IgG (H+L) Donkey Polyclonal Antibody (DyLight® 594)
Catalog # AGRIAS122090
Supplier:  Agrisera

Specifications

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG (H+L)
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 594
  • Flow cytometry:
    Yes
  • Host:
    Donkey
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Rabbit,
    Human,
    Rat
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ambient
  • Immunogen:
    purified rat IgG, whole molecule
  • Purification:
    purified
  • Size:
    1 mg
  • Pk:
    1 mg

Specifications

About this item

Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 594
Epitope:
Host: Donkey
Isotype:
Reactivity: Human, Rabbit, Rat