Anti-PsbA Rabbit Polyclonal Antibody
Catalog # AGRIAS111786
Supplier: Agrisera
Specifications
- Antikörper-Typ:Primär
- Antigen-Symbol:PSBA
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- Reaktivität:A. thaliana,Artemisia annua,Prochlorococcus sp.,Physcomitrella patens,Arundo sp.,Ditylum brightwellii,Pinus strobus,Hordeum vulgare,C. quitensis kunt bartl,Anabaena pcc 7120,Raterostigma sp. Soy bean,Miscanthus giganteus,Spirodela polyrhiza,Panicum miliaceum,N. benthamiana,Lindernia sp.,Symbiodinium sp.,C. reinhardtii,Marchantia polymorpha,Zea mays,Synechococcus sp. pcc 7942,Paulinella chromatophora,Panicum maximum,Coscinodiscus wailesii
- Western blot:Yes
- Epitop:N-terminal
- Kreuzadsorption:No
- UniProtKB:A4QJR4
- Lagertemperatur:–20 °C
- Transporttemperatur:Ambient
- Immunogen:KLH-conjugated synthetic peptide derived from N-terminal of available plant, algal and cyanobacterial PsbA sequences, including Arabidopsis thaliana
- Größe:200 µl
- VE:200 µl
Specifications
About this item
Type: Primary
Antigen: PSBA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Anabaena pcc 7120, A. thaliana, Artemisia annua, Arundo sp., C. reinhardtii, Colobanthus quitensis kunt bartl, Coscinodiscus wailesii, Raterostigma sp. Glycine max, Ditylum brightwellii, Hordeum vulgare, Lindernia sp., Marchantia polymorpha, Miscanthus giganteus, N. benthamiana, Panicum miliaceum, Panicum maximum, Paulinella chromatophora, Physcomitrella patens, Pinus strobus, Prochlorococcus sp., Spirodela polyrhiza, Symbiodinium sp., Synechococcus sp. pcc 7942, Zea mays