Anti-DMPO Mouse Monoclonal Antibody [clone: N1664A]
Catalog # AGRIAS111632
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:DMPO
- Clonality:Monoclonal
- Clone:N1664A
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Multiple
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:5,5-dimethyl-2-(8-octanoic acid)-1-pyrrolone-N-oxide conjugated to ovalbumin
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: DMPO
Clonality: Monoclonal
Clone: N1664A
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Many Species