Order Entry
Switzerland
ContactUsLinkComponent
Anti-SppA1 Rabbit Polyclonal Antibody
Anti-SppA1 Rabbit Polyclonal Antibody
Katalognr. AGRIAS101598
Anbieter:  Agrisera
Anti-SppA1 Rabbit Polyclonal Antibody
Katalognr. AGRIAS101598
Anbieter:  Agrisera

Technische Daten

  • Antibody type:
    Primary
  • Antigen symbol:
    SPPA1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Spinacia oleracea,
    C. reinhardtii,
    Vicia faba,
    A. thaliana,
    Phaseolus vulgaris
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ambient
  • Immunogen:
    recombinant SppA1 protease from Arabidopsis thaliana, NP_565077.2 (Q9C9C0 , At1g73990)
  • Size:
    300 µl
  • Pk:
    300 µl

Technische Daten

Über diesen Artikel

Type: Primary
Antigen: SPPA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, C. reinhardtii, Phaseolus vulgaris, Spinacia oleracea, Vicia faba