Anti-IgG (H+L) Rabbit Polyclonal Antibody (Biotin)
Catalog # AGRIAS101529
Anbieter: Agrisera
Technische Daten
- Antibody type:Secondary
- Antigen symbol:IgG (H+L)
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:+2°C to +8 °C
- Shipping temperature:Ambient
- Immunogen:Purified rat IgG (H&L)
- Purification:purified
- Size:1 mg
- Pk:1 mg
Technische Daten
Über diesen Artikel
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat