Anti-IgG (H+L) Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Catalog # AGRIAS101059
Supplier: Agrisera
Specifications
- Antikörper-Typ:Sekundär
- Antigen-Symbol:IgG (H+L)
- Klonalität:Polyklonal
- Konjugation:TRITC (Tetramethylrhodamine Isothiocyanate)
- ELISA:Yes
- Durchflusszytometrie:Yes
- Wirt:Goat
- ImmunoChemie:Yes
- Reaktivität:Goat
- Kreuzadsorption:No
- Lagertemperatur:+2°C to +8 °C
- Transporttemperatur:Ambient
- Immunogen:Purified Rabbit IgG, whole molecule
- Reinigung:purified
- Größe:500 μg
- VE:0,5 mg
Specifications
About this item
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope:
Host: Goat
Isotype:
Reactivity: Goat