Order Entry
Switzerland
ContactUsLinkComponent
Anti-IgG (H+L) Goat Polyclonal Antibody (Biotin)
Catalog # AGRIAS101023
Anbieter:  Agrisera
Anti-IgG (H+L) Goat Polyclonal Antibody (Biotin)
Catalog # AGRIAS101023
Anbieter:  Agrisera

Technische Daten

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG (H+L)
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Rabbit,
    Goat,
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Storage temperature:
    +2°C to +8 °C
  • Shipping temperature:
    Ambient
  • Immunogen:
    Purified Rat IgG, whole molecule
  • Purification:
    purified
  • Size:
    500 μg
  • Pk:
    0,5 mg

Technische Daten

Über diesen Artikel

Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Goat, Human, Mouse, Rabbit, Rat