Anti-HydA Rabbit Polyclonal Antibody
Catalog # AGRIAS09600
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:HYDA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:A. thaliana
- Western blot:Yes
- Epitope:full length
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:recombinant, full length Chlamydomonas reinhardtii HydA-2 Q8VZZ0
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: HYDA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: full length
Host: Rabbit
Isotype:
Reactivity: A. thaliana