Anti-Oxalate oxidase Rabbit Polyclonal Antibody (Biotin )
AGRIAS09519
: Agrisera
- Antibody type:Primary
- Antigen name:Oxalate oxidase
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Hordeum vulgare
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:native oxalate oxidase have been purified from barley seedlings
- Purification:purified
- Size:1 ml
- Pk:1 mL
Type: Primary
Antigen: Oxalate oxidase
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Hordeum vulgare