Anti-meta-topolin riboside Rabbit Polyclonal Antibody
Catalog # AGRIAS09440
Anbieter: Agrisera
Technische Daten
- Antibody type:Primary
- Antigen name:meta-topolin riboside
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:BSA-conjugated via ribose, meta-topolin riboside
- Tested applications:Immunolocalization
- Size:200 µg
- Pk:200 µG
Technische Daten
Über diesen Artikel
Type: Primary
Antigen: meta-topolin riboside
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: