Anti-Abeta Rabbit Polyclonal Antibody
Catalog # AGRIAS08357
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen name:Amyloid beta 1-42
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Sequence:DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVIA
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:synthetic peptide chosen from human Abeta (1-42) protein. Amino acid sequence: D-A-E-F-R-H-D-S-G-Y-E-V-H-H-Q-K-L-V-F-F-A-E-D-V-G-S-N-K-G-A-I-I-G-L-M-V-G-G-V-V-I-A
- Tested applications:Dot Blot
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Amyloid beta 1-42
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human