Anti-slr1641 Rabbit Polyclonal Antibody
Katalognr. AGRIAS08344
Anbieter: Agrisera
Technische Daten
- Antibody type:Primary
- Antigen symbol:SLR1641
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Synechocystis sp. pcc 6803
- Western blot:Yes
- Epitope:internal
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:recombinant clpB1 protein, derived from Synechocystis PCC 6803 strain slr1641 sequence; protein has an internal translation site. The nomenclature used is reverse of what is mentioned in the cyanobase.
- Size:100 µl
- Pk:100 µl
Technische Daten
Über diesen Artikel
Type: Primary
Antigen: SLR1641
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Synechocystis sp. pcc 6803