Anti-VRN2 Mouse Monoclonal Antibody
AGRIAS08341
: Agrisera
- Antibody type:Primary
- Antigen symbol:VRN2
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:T. aestivum
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide chosen from Triticum aestivum VRN2 sequence
- Size:2 x 50 µg
- Pk:100 µG
Type: Primary
Antigen: VRN2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: T. aestivum