Anti-B-PE Rabbit Polyclonal Antibody
Catalog # AGRIAS08280
Anbieter: Agrisera
Technische Daten
- Antibody type:Primary
- Antigen symbol:B-PE
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:native purified B-phycoerythrin of Porphyridium cruentum with attached chromophores
- Purification:purified
- Size:200 µl
- Pk:200 µl
Technische Daten
Über diesen Artikel
Type: Primary
Antigen: B-PE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human