Order Entry
Switzerland
ContactUsLinkComponent
Anti-GOGAT Rabbit Polyclonal Antibody
Anti-GOGAT Rabbit Polyclonal Antibody
Catalog # AGRIAS07242
Supplier:  Agrisera
undefined
Anti-GOGAT Rabbit Polyclonal Antibody
Catalog # AGRIAS07242
Supplier:  Agrisera

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    GOGAT
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Miscanthus giganteus,
    Zea mays,
    A. thaliana,
    Spartina sp.,
    Phaseolus vulgaris
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ambient
  • Immunogen:
    KLH-conjugated synthetic peptide well conserved in known GOGAT sequences from different species including Arabidopsis thaliana Fd-GOGAT 1 Q9ZNZ7, At5g04140 and Fd-GOGAT 2 Q9T0P4, At2g41220
  • Size:
    200 µl
  • Pk:
    50 µl

Specifications

About this item

Type: Primary
Antigen: GOGAT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, Miscanthus giganteus, Phaseolus vulgaris, Spartina sp., Zea mays