Order Entry
Switzerland
ContactUsLinkComponent
Anti-HYL1 Rabbit Polyclonal Antibody
Anti-HYL1 Rabbit Polyclonal Antibody
Catalog # AGRIAS06136
Supplier:  Agrisera
Anti-HYL1 Rabbit Polyclonal Antibody
Catalog # AGRIAS06136
Supplier:  Agrisera

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    HYL1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Chlamydomonas reinhardtii
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ambient
  • Immunogen:
    KLH-conjugated synthetic peptide derived from Arabidopsis thaliana Hyl1 protein sequence O04492, At1g09700
  • Size:
    200 µl
  • Pk:
    200 µl

Specifications

About this item

Type: Primary
Antigen: HYL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chlamydomonas reinhardtii