Anti-PsaF Rabbit Polyclonal Antibody
Catalog # AGRIAS06104
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:PSAF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Oryza sativa,Hordeum vulgare
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide derived from the PsaF protein sequence of Arabidopsis thaliana (At1g31330). This peptide sequence is not completely conserved in mono- and dicots.
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: PSAF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Hordeum vulgare, Oryza sativa