Anti-PsbX Rabbit Polyclonal Antibody
N° de catalogue AGRIAS05066
Fournisseur: Agrisera
Spécifications
- Antibody type:Primary
- Antigen symbol:PSBX
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Spinacia oleracea,Ricinus communis,Zea mays,A. thaliana,N. tabacum,H. vulgare
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide dervied from Arabidopsis thaliana PsbX sequence At2g06520
- Size:200 µl
- Pk:200 µl
Spécifications
À propos de cet article
Type: Primary
Antigen: PSBX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, H. vulgare, N. tabacum, Ricinus communis, Spinacia oleracea, Zea mays