Order Entry
Switzerland
ContactUsLinkComponent
Anti-AOX1/2 Rabbit Polyclonal Antibody
Catalog # AGRIAS04054
Anbieter:  Agrisera
Anti-AOX1/2 Rabbit Polyclonal Antibody
Catalog # AGRIAS04054
Anbieter:  Agrisera

Technische Daten

  • Antibody type:
    Primary
  • Antigen name:
    Aldehyde Oxidase 1/2
  • Antigen symbol:
    AOX1/2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Cross adsorption:
    No
  • UniProtKB:
    Q7XT33,
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ambient
  • Immunogen:
    KLH-conjugated synthetic peptide derived from fully conserved C-terminal consensus motif from plant AOX isoforms 1 and 2 including Arabidopsis thaliana AOX1A
  • Size:
    200 µl
  • Pk:
    50 µl

Technische Daten

Über diesen Artikel

Type: Primary
Antigen: AOX1/2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human