Anti-RPN2 Rabbit Polyclonal Antibody
Catalog # ORIGTA590669
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:RPN2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Concentrated
- Storage buffer:20 mM Potassium Phosphate, 150 mM Sodium Chloride, pH 7.0
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:DNA immunization. This antibody is specific for the Middle Region of the target protein.
- Purification:The fragment used for DNA immunization was expressed in E.coli and the purified protein fragment was used for affinity purification of the antibody.
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1:500; ELISA: 1:100-1:2000; IHC: 1:100
Type: Primary
Antigen: RPN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human