Order Entry
Switzerland
ContactUsLinkComponent
Anti-ANO1 Mouse Monoclonal Antibody [clone: 5H2]
Catalog # ORIGTA803579
Anbieter:  OriGene
Anti-ANO1 Mouse Monoclonal Antibody [clone: 5H2]
Catalog # ORIGTA803579
Anbieter:  OriGene

Technische Daten

  • Antibody type:
    Primary
  • Antigen name:
    Anoctamin 1, calcium activated chloride channel
  • Antigen symbol:
    ANO1
  • Clonality:
    Monoclonal
  • Clone:
    5H2
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Antigen synonyms:
    TMEM16A
  • Amino acid number:
    1 to 333
  • Storage buffer:
    PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Human recombinant protein fragment corresponding to amino acids 1-333 of human ANO1 (NP_060513) produced in E.coli.
  • Purification:
    purified by affinity chromatography
  • Size:
    100 μl
  • Pk:
    100 µl

Technische Daten

Über diesen Artikel

Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: ANO1
Clonality: Monoclonal
Clone: 5H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human