Anti-PLA2G6 Mouse Monoclonal Antibody [clone: 10A2]
ORIGTA803198
: OriGene
- Antibody type:Primary
- Antigen symbol:PLA2G6
- Clonality:Monoclonal
- Clone:10A2
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Amino acid number:490 to 752
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:83.9 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 490-752 of human PLA2G6 (NP_001004426) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μl
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:500
Type: Primary
Antigen: PLA2G6
Clonality: Monoclonal
Clone: 10A2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human