Anti-Hcn3 Rabbit Polyclonal Antibody
Catalog # ORIGTA328956
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:HCN3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Rat
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:727 to 744
- Storage buffer:Lyophilized. Concentration before lyophilization ~0,8mg/ml (lot dependent, please refer to CoA along with shipment for actual concentration). Buffer before lyophilization: Phosphate buffered saline (PBS), pH 7,4, 1% BSA, 0,05% NaN3.
- Sequence:C-QRATGDGSPRRKGSGSER
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Peptide (C)QRATGDGSPRRKGSGSER corresponding to amino acid residues 727-744 of rat HCN3 . Intracellular, C-Terminus.
- Purification:purified by antigen affinity chromatography
- Size:200 μl
- Pk:200 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:200-1:2000; Immunochemistry: 1:100-1:3000
Type: Primary
Antigen: Hcn3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat