Anti-CRTC2 Rabbit Polyclonal Antibody
ORIGTA320138
: OriGene
- Antibody type:Primary
- Antigen symbol:CRTC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Storage buffer:CRTC2 Antibody is supplied in PBS containing 0,02% sodium azide.
- Molecular weight:Predicted: 67, 76 kDa; Observed: 70 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:CRTC2 antibody was raised against a 16 amino acid peptide near the carboxy terminus of human CRTC2.
- Purification:purified by affinity chromatography via peptide column
- Size:100 μg
- Pk:100 µG
Recommended Dilutions: Western Blot: 1-2 ?g/ml; IHC; Immunofluorescence
Type: Primary
Antigen: CRTC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat