Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-FGF21 Mouse Monoclonal Antibody [clone: 2E2]
  ORIGCF502954
 :  OriGene
Anti-FGF21 Mouse Monoclonal Antibody [clone: 2E2]
  ORIGCF502954
 :  OriGene

 

  • Antibody type:
    Primary
  • Antigen symbol:
    FGF21
  • Clonality:
    Monoclonal
  • Clone:
    2E2
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Amino acid number:
    29 to 209
  • Storage buffer:
    PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Molecular weight:
    19.4 kDa
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Human recombinant protein fragment corresponding to amino acids 29-209 of human FGF21(NP_061986) produced in E.coli.
  • Purification:
    purified by affinity chromatography
  • Size:
    100 μg
  • Pk:
    100 µG

 

 

Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: FGF21
Clonality: Monoclonal
Clone: 2E2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human