Anti-Mapk8ip1 Rabbit Polyclonal Antibody
Catalog # ORIGTA318944
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:MAPK8IP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Storage buffer:100µg (0,2mg/ml) protein A purified rabbit anti-JIP1 polyclonal antibody in phosphate-buffered saline (PBS) containing 0,5% BSA, 30% glycerol, and 0,01% thimerosal.
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Synthetic peptide from mouse JIP1
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 0.5-4?g/ml
Type: Primary
Antigen: Mapk8ip1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat