Baths and lids, digital, stainless steel
Anbieter: witeg Labortechnik
Zertifikate
Über diesen Artikel
These stainless steel baths are ideal for biotechnology, clinical, environmental, medical, petroleum, pharmaceutical or industrial applications.
- Stainless steel lid prevents evaporation and keeps temperature constant
- Durability and high thermal efficiency
- Digital fuzzy controller with jog - shuttle switch
- Digital LCD display
Frequently Bought Together