Sample bottles, narrow neck, round, HDPE, Boston type
Supplier: EP SCIENTIFIC
Certificates
About this item
HDPE container with white LDPE-lined polypropylene cap.
- Narrow mouth for precision pouring
- Caps assembled onto containers
- Certified to meet or exceed EPA performance-based standards for metals, cyanide and fluoride
- Certificate of Analysis (CofA) included in the case and barcoded with lot number for traceability
- Each container individually barcoded with lot number and unique container number
- Analysis labels included unattached for optional use
- Carton sealed with security tape providing tamper evidence
Recommended for collecting water samples and other low viscosity liquids to be analysed for inorganic trace analytes.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
- Catalog No:
- EPSC319-1000
- EPSC319-0125
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement