Sampling bottles, narrow/wide neck, HDPE
HDPE, with leak-proof PP/HDPE closure.
- Narrow mouth for easy handling and precision pouring
- Wide mouth for easy aseptic filling
- Leakproof and break resistant
- Certified to meet or exceed EPA performance-based standards for metals, cyanide and fluoride
- Each container individually barcoded with lot number and unique container number
Recommended for water and other environmental liquid sample collections, especially for critical projects where sample containment and transport is of elevated concern.
: Certificate of analysis (CofA) included in the case and barcoded with lot number for traceability. Analysis labels included unattached for optional use.
HDPE, with leak-proof PP/HDPE closure.
- Narrow mouth for easy handling and precision pouring
- Wide mouth for easy aseptic filling
- Leakproof and break resistant
- Certified to meet or exceed EPA performance-based standards for metals, cyanide and fluoride
- Each container individually barcoded with lot number and unique container number
Recommended for water and other environmental liquid sample collections, especially for critical projects where sample containment and transport is of elevated concern.
: Certificate of analysis (CofA) included in the case and barcoded with lot number for traceability. Analysis labels included unattached for optional use.
- Catalog No:
- EPSCN319-0125
- EPSCN319-0250
- EPSCN319-0500
- EPSCN319-1000
- EPSCN311-0125
- EPSCN311-0125BPC
- EPSCN311-0250
- EPSCN311-0250BPC
- EPSCN311-0500
- EPSCN311-0500BPC
- EPSCN311-1000
- EPSCN311-1000BPC
- EPSCN301-0125
- EPSCN301-0250
- EPSCN301-0500
- 216-4433
- 216-4434
- EPSCN411-0500
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
Product Family Options
Narrow neck
- CapacityDescriptionColourHeightExt.ØCap colourPacked
- 125 mlCertified metals and water quality parametersNatural101 mm50 mmNatural72/case1 * 72 STSFr. 388.00
Low Manufacturing Impact Specifications
Cat. No.EPSCN319-0125Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationIRECS; ISO 14001 (Certificate Registration No. EMS 752603)Low Manufacturing Impact - 250 mlCertified metals and water quality parametersNatural133 mm61 mmWhite72/case1 * 72 STSFr. 482.00
Low Manufacturing Impact Specifications
Cat. No.EPSCN319-0250Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - 500 mlCertified metals and water quality parametersNatural171 mm73 mmNatural48/case1 * 48 STSFr. 387.00
Low Manufacturing Impact Specifications
Cat. No.EPSCN319-0500Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - 1000 mlCertified metals and water quality parametersNatural217 mm92 mmNatural24/case1 * 24 STSFr. 266.00
Low Manufacturing Impact Specifications
Cat. No.EPSCN319-1000Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact
Wide neck
- CapacityDescriptionColourHeightExt.ØCap colourPacked
- 125 mlCertified for metals and water quality parametersNatural99 mm50 mmNatural72/case1 * 72 STSFr. 392.00
Low Manufacturing Impact Specifications
Cat. No.EPSCN311-0125Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationIRECS; ISO 14001 (Certificate Registration No. EMS 752603)Low Manufacturing Impact - 125 mlCertified for metals and water quality parametersNatural99 mm50 mmNatural500/case1 * 500 STSFr. 2’250.00
Low Manufacturing Impact Specifications
Cat. No.EPSCN311-0125BPCEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - 250 mlCertified for metals and water quality parametersNatural133 mm61 mmNatural72/case1 * 72 STSFr. 490.00
Low Manufacturing Impact Specifications
Cat. No.EPSCN311-0250Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - 250 mlCertified for metals and water quality parametersNatural133 mm61 mmNatural250/case1 * 250 STSFr. 1’500.00
Low Manufacturing Impact Specifications
Cat. No.EPSCN311-0250BPCEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - 500 mlCertified for metals and water quality parametersNatural168 mm73 mmNatural48/case1 * 48 STSFr. 392.00
Low Manufacturing Impact Specifications
Cat. No.EPSCN311-0500Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - 500 mlCertified for metals and water quality parametersNatural168 mm73 mmNatural125/case1 * 125 STSFr. 1’000.00
Low Manufacturing Impact Specifications
Cat. No.EPSCN311-0500BPCEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - 1000 mlCertified for metals and water quality parametersNatural199 mm91 mmNatural24/case1 * 24 STSFr. 293.00
Low Manufacturing Impact Specifications
Cat. No.EPSCN311-1000Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - 1000 mlCertified for metals and water quality parametersNatural199 mm91 mmNatural50/case1 * 50 STSFr. 574.00
Low Manufacturing Impact Specifications
Cat. No.EPSCN311-1000BPCEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - 125 mlCertified metals and water quality parameters, Diquot/ParaquotAmber99 mm50 mmAmber72/case1 * 72 STSFr. 392.00
Low Manufacturing Impact Specifications
Cat. No.EPSCN301-0125Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationIRECS; ISO 14001 (Certificate Registration No. EMS 752603)Low Manufacturing Impact - 250 mlCertified metals and water quality parameters, Diquot/ParaquotAmber133 mm61 mmAmber72/case1 * 72 STSFr. 490.00
Low Manufacturing Impact Specifications
Cat. No.EPSCN301-0250Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationIRECS; ISO 14001 (Certificate Registration No. EMS 752603)Low Manufacturing Impact - 500 mlCertified metals and water quality parameters, Diquot/ParaquotAmber168 mm73 mmAmber48/case1 * 48 STSFr. 392.00
Low Manufacturing Impact Specifications
Cat. No.EPSCN301-0500Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - 125 mlCertified sterile, metals and water quality, Cyanide, FluorideNatural99 mm50 mmWhite48/case1 * 48 STSFr. 382.00
Low Manufacturing Impact Specifications
Cat. No.216-4433Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of Retirement - 250 mlCertified sterile, metals and water quality, Cyanide, FluorideNatural133 mm61 mmWhite60/case1 * 60 STSFr. 582.00
Low Manufacturing Impact Specifications
Cat. No.216-4434Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of Retirement - 500 mlCertified sterile, metals and water quality, Cyanide, FluorideNatural168 mm73 mmWhite40/case1 * 40 STSFr. 509.00
Low Manufacturing Impact Specifications
Cat. No.EPSCN411-0500Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of Retirement