ContactUsLinkComponent
Qualitative filter papers, slow filtration
Qualitative filter papers, slow filtration
Supplier:  PRAT DUMAS
Certificates
undefined
Qualitative filter papers, slow filtration
Supplier:  PRAT DUMAS
Specifications
  • Catalog No:
  • PRATA015210
  • PRATA007210
  • PRATJ015210
  • PRATA009210
  • Weight:
  • 100 g/m²
  • 100 g/m²
  • 100 g/m2
  • 100 g/m²
  • Thickness:
  • 200 µm
  • 200 µm
  • 200 µm
  • 200 µm
  • Particle retention:
  • 2 - 3 µm
  • 2 - 3 µm
  • 2 - 3 µm
  • 2 - 3 µm