ContactUsLinkComponent
Digital camera, Axiocam MRc
Catalog # ZEIS4265089901000
Supplier:  ZEISS
undefined
Digital camera, Axiocam MRc
Catalog # ZEIS4265089901000
Supplier:  ZEISS
Specifications
  • Model:
    AxioCam MRc
  • Resolution:
    1,4 MP
  • Sensor type:
    CCD
  • Sensor size:
    2/3"
  • Description:
    Digital camera
  • Environmentally Preferable:
  • Pixel size:
    6,45×6,45 µm
  • Pixels (W×H):
    1388×1040
  • Operating temperature:
    5...35 °C
  • Pk:
    1
Specifications
About this item
High resolution camera made of blue anodized aluminum with cooling fins.
  • One stage Peltier cooling
  • Readout of sensor sub-regions (ROI)
  • FireWire / IEEE1394 including cable and IR barrier filter BG40
  • TTL control for controlling of external shutters
  • Mid-range colour driver