Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-SLC12A1 Rabbit Polyclonal Antibody
  BSBTPB9392
undefined
Anti-SLC12A1 Rabbit Polyclonal Antibody
  BSBTPB9392

 

  • Antibody type:
    Primary
  • Antigen name:
    solute carrier family 12 (sodium/potassium/chloride transporter), member 1
  • Antigen symbol:
    Solute carrier family 12, member 1
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Gene ID:
    6557
  • Antigen synonyms:
    BSC1|NKCC2
  • UniProtKB:
    Q13621
  • Storage temperature:
    At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the N-terminus of human SLC12A1(52-83aa DEAQKRLRISFRPGNQECYDNFLQSGETAKTD), different from the related mouse sequence by two amino acids, and from the related rat sequence by four amino acids.
  • Purification:
    Immunogen affinity purified.
  • Size:
    100 µg/vial
  • Pk:
    100 µG

 

 

Rabbit IgG polyclonal antibody for Solute carrier family 12 member 1(SLC12A1) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Solute carrier family 12 (sodium/potassium/chloride transporters), member 1, also called NKCC2 is specifically found in cells of the thick ascending limb of the loop of Henle in nephrons, the basic functional units of the kidney. This gene is mapped to 15q21.1. This gene encodes a kidney-specific sodium-potassium-chloride cotransporter that is expressed on the luminal membrane of renal epithelial cells of the thick ascending limb of Henle's loop and the macula densa. It plays a key role in concentrating urine and accounts for most of the NaCl resorption. It is sensitive to such diuretics as furosemide and bumetanide. Some Bartter-like syndromes result from defects in this gene. This gene plays a vital role in the regulation of ionic balance and cell volume.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.

WB: The detection limit for SLC12A1 is approximately 0.1ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: SLC15A2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
, Human