ICR swab
Supplier: MERCK LIFE SCIENCE
Certificates
About this item
The ideal swab for microbial monitoring of isolator and clean room surfaces – Requires opening only once for sampling. The ICR Swab is a unique system for environmental monitoring of dry surfaces, such as filling needles, equipment recesses, uneven surfaces and tubing in clean rooms and isolators. It is a qualitative presence/absence test for microbiological contamination. This ‘all-in-one’ heipha ICR Swab System requires opening only once for sampling. All further steps, such as addition of broth, incubation and detection of turbidity, are performed within the closed system, thus minimizing the risk of potential contaminations due to handling.
- Secure and reliable
- Convenient
- Flexible
- Gamma-irradiated
- Triple-sleeved
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Frequently Bought Together