Orders LinkContactUsLinkComponent
Replacement impeller for Magna Flex™ flasks
Replacement impeller for Magna Flex™ flasks
Supplier:   DWK Life Sciences
Replacement impeller for Magna Flex™ flasks
Supplier:  DWK Life Sciences
Certificates
Replacement impeller for Magna Flex™ flasks
Replacement impeller for Magna Flex™ flasks
4 Options Available Below
Product Details & Documents
This unit includes a top cap with liners, glass stirring ball and connecting silicone tubing. This glass impeller assembly is designed for replacement on all Magna Flex™ style flasks.
Product Family Options
Product Information
  • Availability
    Price
  • 734-3143
    Replacement impeller for 125 ml Magna Flex® spinner flask
    Replacement impeller for 125 ml Magna Flex® spinner flask
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 304.00
    SFr. 304.00
    1 * 1 ST
       SFr. 304.00
  • 734-3144
    Replacement impeller for 250 ml Magna Flex® spinner flask
    Replacement impeller for 250 ml Magna Flex® spinner flask
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 312.00
    SFr. 312.00
    1 * 1 ST
       SFr. 312.00
  • 734-3145
    Replacement impeller for 500 ml Magna Flex® spinner flask
    Replacement impeller for 500 ml Magna Flex® spinner flask
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 415.00
    SFr. 415.00
    1 * 1 ST
       SFr. 415.00
  • 734-3149
    Replacement impeller for 8000 ml Magna Flex® spinner flask
    Replacement impeller for 8000 ml Magna Flex® spinner flask
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 370.00
    SFr. 370.00
    1 * 1 ST
       SFr. 370.00